EMAIL THIS PAGE TO A FRIEND
S1023 Sigma

Silicon nitride

predominantly β-phase

Purchase

Properties

InChI Key   HQVNEWCFYHHQES-UHFFFAOYSA-N
density   3.44 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?