EMAIL THIS PAGE TO A FRIEND
S5629 Sigma-Aldrich

Sodium chloride

suitable for plant cell culture, ≥99.5%

Synonym: Halite

Purchase

Properties

assay   ≥99.5%
application(s)   cell culture | plant: suitable
mp   801 °C (lit.)
SMILES string   [Na+].[Cl-]
InChI   1S/ClH.Na/h1H;/q;+1/p-1
InChI key   FAPWRFPIFSIZLT-UHFFFAOYSA-M

Description

Other Notes

Easily compare specifications for Sodium Chloride products with the Sodium Chloride specification table.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
VZ4725000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?