• USA Home
  • SIAL0008 - Sigma® cell lifter
EMAIL THIS PAGE TO A FRIEND
SIAL0008 Sigma-Aldrich
Sigma® cell lifter
blade L 1.9 cm, handle L 18 cm, individually wrapped, sterile, polyethylene, disposable
  •  NACRES NB.22
Purchase
Properties
Related Categories Cell Culture Supplies, Labware, Sigma Cell Lifter and Cell Scraper, Sigma Cultureware
material   polyethylene
sterility   sterile
feature   disposable
  individually wrapped
blade L   1.9 cm
handle L   18 cm
suitability   suitable for (harvesting stem cells and colonies while maintaining high viability)
Description
General description
• Used for scraping cells from dishes and plates
• Ideal tool for harvesting stem cells and colonies while maintaining high viability
• One piece design for better control
• Blade design minimizes cell damage and ensures even contact with the growth surface
• Individually wrapped
• Sterilized by gamma radiation and certified nonpyrogenic
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?