EMAIL THIS PAGE TO A FRIEND
SML1803 Sigma-Aldrich

NVS-CECR2-1

≥98% (HPLC)

Synonym: Cyclopropyl-{2-(propane-1-sulfonyl)-6-[1-(2,2,6,6-tetramethyl-piperidin-4-yl)-1H-indol-5-yl]-pyrimidin-4-yl}-amine, NVS-1

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Gene Regulation,
Quality Level   100
assay   ≥98% (HPLC)
form   powder
color   white to beige
solubility   DMSO: 10 mg/mL, clear
storage temp.   −20°C
SMILES string   CC(NC(C)(C)C1)(C)CC1N2C=CC3=C2C=CC(C4=NC(S(CCC)(=O)=O)=NC(NC5CC5)=C4)=C3

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

NVS-CECR2-1 (NVS-1) is a potent and selective inhibitor of CECR2 (cat eye syndrome chromosome region, candidate 2) gene. CECR2 is duplicated in the human disorder cat eye syndrome, which includes defects of the eye, heart, anus, kidney, skeleton, face, and may include mild mental deficiency. CECR2 is involved in neurulation, the folding process that results in the transformation of the neural plate into the neural tube, and is also thought to play a role in DNA damage response. NVS-CECR2-1 (NVS-1) binds to CECR2 with high affinity. It has an IC50 value of 47 nM in Alpha screen and a KD of 80 nM in ITC. NVS-CECR2-1 showed no cross reactivity in a BRD panel of 48 targets or any major activity in kinase, protease and receptor panels. For full characterization details, please visit the NVS-CECR2-1 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

NVS-CECR2-1 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Discover Bioactive Small Molecules for Gene Regulation

The loss of regulation of gene expression is a key component to many human disease states, including neurodegenerative disorders, autoimmune conditions and, most prominently, cancers. Regulation of g...
Keywords: Acetylations, Epigenetics, Gene expression, Genomics, Methylations, PAGE, Post translational modifications, Transcription

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?