• USA Home
  • Z370258 - Enzyme Immunoassays: From Concept to Product Development

EMAIL THIS PAGE TO A FRIEND
Z370258 Sigma

Enzyme Immunoassays: From Concept to Product Development

  •  ISBN-10 0-412-05601-1

  •  ISBN-13 978-0-412-05601-7

Purchase

Properties

publication info   S. Deshpande, Kluwer Academic, 1996, 288 pp., hard cover
mfr. no.   Kluwer Academic Publishers

Description

General description

A vertical exposition of enzyme immunoassays; the intended audience is product designers and R&Dgroups who aim to commercialize EIA technology. From concepts of antibody and antigen through laboratory development, all the way to manufacturing, marketing, and regulatory compliance, the book is a practical resource for the diagnostic field.

Table of Contents

Part 1: Basic considerations (concepts): Introduction;
Antibody: biochemistry, structure, and function;
Antigen: antibody reactions;
Conjugation techniques;
Antibody production;
Enzymes and signal amplification systems;
Separation and solid phase systems;
Part 2: Product development: Immunoassay classification and commercial technologies;
Assay development, evaluation, and validation;
Reagent formulations and shelf life evaluation;
Data analysis;
Documentation, Registration, and Diagnostic Start-Ups.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?