EMAIL THIS PAGE TO A FRIEND
Z373672 Sigma
Immunoassay
  •  ISBN-10 0-12-214730-8
  •  ISBN-13 978-0-12-214730-2
Purchase
Properties
publication info   E. Diamandis and T. Christopoulus, Academic Press, San Diego, CA, 1996, 579 pp., soft cover
mfr. no.   Elsevier
Description
General description
An indispensable guide to information on the theory and practice of immunoassays. This book balances the content among theory, practical issues, quality control, automation, and subspecialty areas. Protocols for specific assays are included.
Table of Contents
Preface. Past, Present, and Future of Immunological Assays. Immune Function and Antibody Structure. Theory of Immunoassays. Data Interpretation and Quality Control. , Production and Purification of Antibodies. Antibody Engineering. Interferences in Immunoassays. Labeling of Antibodies and Antigens. Solid-Phases in Immunoassay.
Immunoassay Configurations. The Avidin–Biotin System. Radioimmunoassay. Enzyme Immunoassay. Fluorescence Immunoassays. Chemiluminescence Immunoassay. Bioluminescence Immunoassays. Nephelometric and Turbidimetric Immunoassay. Simultaneous Multianalyte
Immunoassays. Noncompetitive Immunoassay for Small Molecules. Free Hormone Measurements. Automation of Immunological Assays. Thin-Film Immunoassays. Immunoblotting Techniques. Development of In-House Immunological Assays. Chapter References. Subject Index.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?