• USA Home
  • Z617652 - Greiner CrystalQuick® 96 well plates for protein crystallization

EMAIL THIS PAGE TO A FRIEND
Z617652

Greiner CrystalQuick® 96 well plates for protein crystallization

CrystalDrop plate lid

Synonym: 96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Purchase

Properties

material   polystyrene
sterility   non-sterile
mfr. no.   Greiner 609150
packaging   case of 80
L × W × H   127.35 mm × 84.8 mm × 6.0 mm
color   clear lid
matrix active group   non-treated surface treatment

Description

General description

• Plate is optimized for the sitting drop application
• 96 reservoirs with flat bottom, 320 μL each
• 288 round crystallisation wells, 1.9 μL each
• Hanging drop application possible in combination with the CrystalDrop lid

CrystalDrop Lid for CrystalQuick® 96 well Plates.

Lid for CrystalQuick 96 well microplates.
• enables individual sealing of the 96 wells
• 2 hanging drop positions per well
• Wells with a diam. 2.7 mm for the samples

Suitability

Lid enables sealing of 96 Well CrystalQuick(tm) Plates, 2 wells per reservoir and printing mask, for hanging drop applications

Legal Information

CrystalDrop is a trademark of Greiner Bio-One GmbH

CrystalQuick is a registered trademark of Greiner Bio-One GmbH

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?