• USA Home
  • Z709417 - Corning® 384 well plate accessories
EMAIL THIS PAGE TO A FRIEND
Z709417
Corning® 384 well plate accessories
Sealing tape, 384 well
Purchase
Properties
material   (optical polymer)
mfr. no.   Corning 3775
Description
General description
The 384 well Corning Crystallization Plate is designed for full automation in crystal screening. It was built to meet standard 384 well microplate footprint and well locations.
• It features 192 large reservoir wells and corresponding protein wells.
• Reservoir (reagent) wells and protein wells are positioned to be compatible with multi-head dispenser (up to 96 well heads).
• It is made of an advanced optical polymer superior to polypropylene and polystyrene, which has high optical clarity and greater resistance to organic solvents.
Legal Information
Corning is a registered trademark of Corning, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?