• USA Home
  • Z740078 - BRAND® pipette tips low retention, racked, TipBox

EMAIL THIS PAGE TO A FRIEND
Z740078

BRAND® pipette tips low retention, racked, TipBox

volume 5-300 μL, non-sterile, pack of 480 ea (5 boxes of 96)

  •  NACRES NB.01

Purchase

Properties

Related Categories BRAND, BRAND Low Retention Pipette Tips, BRAND Pipette Tips, BRAND Pipettes, Pipettors, and Tips, Labware,
material   colorless
  colorless polypropylene tip
  polypropylene
sterility   non-sterile
feature   RNase and DNase free
  endotoxin free
packaging   pack of 480 ea (5 boxes of 96)
mfr. no.   BRAND, 732310
maximum volume   300 μL
volume   5-300 μL
binding type   low binding surface

Description

General description

Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.

Quality features:
• High-purity polypropylene, free from DiHEMDA and oleamide
• Manufactured without lubricants; cadmium-free pigments
• Graduation for a quick volume check
• New: All pipette tips and filter tips up to 1000 μL are free of DNA (<40 fg), RNase (<8.6 fg), endotoxins (<1 pg) and ATP (<1 fg)
• New: All sterile tips and packaging are manufactured under BIO-CERT® quality certification: Sterile acc. ISO 11 137 and AAMI guidelines, a SAL 10-6 is obtained
• Supplied with Certificate of analysis
• New: TipBox/Rack System: all sizes up to 1000 μL available in 96-unit format
• Autoclavable at 121°C (2 bar), acc. DIN EN 285
• Environmentally friendly packaging systems
• Pipette and filter tips from BRAND are tested for BRAND pipettes and most of the pipette types of Gilson®, Thermo Fisher Scientific Finnpipette®, Eppendorf® and Biohit®/Sartorious
• The 5 mL tip is exclusively tested for BRAND pipettes and Thermo Fisher Scientific Finnpipette®
• The 10 mL tip is suitable for BRAND, Eppendorf® and Gilson®

Ultra Low Retention-Pipette tips and Filter tips (ULR)The surfaces of the Ultra Low Retention tips are produced through a special, patented physicochemical process. The homogeneous, defect-free surface thus produced has extremely low surface tension - over 50% less than PTFE (see table). This significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.

• Ideal for biological samples that contain detergents such as Triton X-100, SDS, Tween etc
• No additives that can be leached out! No siliconization of the surface!
• High chemical resistance. Ideal for working with solvents
• The tips can be autoclaved at 121°C (2 bar) without damaging the material properties

Other Notes

BR732310 (India)

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG

BRAND is a registered trademark of BRAND GMBH + CO KG

Biohit is a registered trademark of Sartorius Biohit Liquid Handling Oy

Eppendorf is a registered trademark of Eppendorf AG

Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Gilson is a registered trademark of Gilson, Inc.

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

BRAND® Ultra Low Retention Tips - Comparison of Various Competitor Products Using Photometric Measurements

The wetting of pipette tips when pipetting non-aqueous liquids plays a critical role in the life sciences. Liquids, such as protein solutions, liquids which contain detergents or glycerin, wet the su...
Keywords: Detergents, Polymerase chain reaction

Determining Residual Liquids Using Gravimetric Measurement Using BRAND® Ultra Low Retention Tips

The moistening of untreated standard tips while pipetting detergent-containing or viscous media is a general problem in the laboratory. A film forms on the surface of the polypropylene that, when del...
Rainer Feldmann, Kathrin Kraft, Lisa Pollak, Thorsten Rawinski
Keywords: Detergents, Electrophoresis, Gel electrophoresis, PAGE, Polymerase chain reaction

Related Content

Ultra Low Retention Pipette Tips from BrandTech® Scientific

Thanks to the patented treatment, the surface of the BrandTech® Scientific Ultra Low Retention Pipette Tips are ultra-hydrophobic. Special patented process does not use coatings that might lead to sa...
Keywords: Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?