• USA Home
  • A224 - (+)-Anatoxin-A fumarate salt

EMAIL THIS PAGE TO A FRIEND
A224 Sigma

(+)-Anatoxin-A fumarate salt

solid

Synonym: (+)-AnTx-a, (+)-2-Acetyl-9-azabicyclo[4.2.1]non-2-ene fumarate salt, (1R)-1-(9-Azabicyclo[4.2.1]non-2-en-2-yl)ethanone fumarate salt

  • CAS Number 64285-06-9

  • Empirical Formula (Hill Notation) C10H15NO · C4H4O4

  • Molecular Weight 281.30

  •  MDL number MFCD00274024

Purchase

Properties

form   solid
optical activity   [α]20/D +25.1°, c = 0.49 in H2O(lit.)
color   light brown
solubility   H2O: 15 mg/mL
storage temp.   2-8°C

Description

Biochem/physiol Actions

Potent stereospecific nicotinic acetylcholine receptor agonist produced by several toxigenic strains of cyanobacteria.

Caution

Hygroscopic, store desiccated

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
KM5527000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?