• USA Home
  • 29509-U - SLB®-IL60 Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
29509-U Supelco

SLB®-IL60 Capillary GC Column

L × I.D. 60 m × 0.32 mm, df 0.26 μm

Purchase

Properties

Related Categories All Gas Chromatography Columns, Analytical/Chromatography, Capillary GC Columns, Capillary GC Columns, by name, Gas Chromatography (GC),
material   fused silica
parameter   35-300 °C temperature (isothermal or programmed)
Beta value   313
df   0.26 μm
application(s)   gas chromatography (GC): suitable
  gas chromatography (GC): suitable
L × I.D.   60 m × 0.32 mm
matrix active group   Non-bonded; 1,12-Di(tripropylphosphonium)dodecane bis(trifluoromethanesulfonyl)imide phase
Featured Industry   Agriculture
Biofuels
Chemicals and Industrial Polymers
Cleaning Products
Clinical
Cosmetics
Environmental
Flavors and Fragrances
Food and Beverages
Personal Care
Petroleum
column type   capillary polar

Description

General description

Application: Modified (deactivated) version of SLB-IL59 provides better inertness. Selectivity more polar than PEG/wax phases, resulting in unique elution patterns. Higher maximum temperature than PEG/wax columns (300 ºC compared to 270-280 ºC). Excellent alternative to existing PEG/wax columns. Also a good GCxGC column choice. Launched in 2012.
USP Code: None
Phase:
• Non-bonded
• 1,12-Di(tripropylphosphonium)dodecane bis(trifluoromethylsulfonyl)imide
Temp. Limits:
• 35 °C to 300 °C (isothermal or programmed)

Legal Information

SLB is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Supelco SLB-IL60 Ionic Liquid GC Columns: Better High Temperature Stability

The SLB®-IL60 gas chromatography (GC) column is based on an ionic liquid stationary phase platform and displays desirable features that existing non-ionic liquid columns do not. This is the third of ...
Leonard M. Sidisky, Katherine K. Stenerson, Michael D. Buchanan
Reporter US Volume 33.2
Keywords: Chromatography, Degradations, Flame ionization detector, Gas chromatography, Indicators, Size-exclusion chromatography, Solvents

Supelco SLB-IL60 Ionic Liquid GC Columns: Improved Resolution

The SLB®-IL60 gas chromatography (GC) column is based on an ionic liquid stationary phase platform and displays desirable features that existing non-ionic liquid columns do not. This is the second of...
Leonard M. Sidisky, Katherine K. Stenerson, Michael D. Buchanan
Reporter US Volume 33.1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography, Solvents

Supelco SLB-IL60 Ionic Liquid GC Columns: Lower FID Bleed

The SLB®-IL60 gas chromatography (GC) column is based on an ionic liquid stationary phase platform, and displays desirable features that existing non-ionic liquid columns do not. This is the fourth o...
Leonard M. Sidisky, Katherine K. Stenerson, and Michael D. Buchanan
Reporter US Volume 33.3
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Phase transitions, Solvents

Supelco SLB-IL60 Ionic Liquid GC Columns: Stability to Loss of Carrier Gas Flow

The SLB®-IL60 gas chromatography (GC) column is based on an ionic liquid stationary phase platform, and displays desirable features that existing non-ionic liquid columns do not. This is the fifth of...
Leonard M Sidisky, Katharine K Stenerson, Michael D Buchanan
Reporter US Volume 33.4
Keywords: Chromatography, Degradations, Flame ionization detector, Gas chromatography, Size-exclusion chromatography, Solvents

Supelco SLB-IL60 Ionic Liquid GC Columns: Unique Selectivity

The SLB®-IL60 gas chromatography (GC) column is based on the ionic liquid stationary phase platform, and displays desirable features that existing non-ionic liquid columns do not. This is the first o...
Leonard M. Sidisky, Katherine K. Stenerson, Michael D. Buchanan
Reporter US Volume 32.2
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography, Solvents

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors:
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?