• USA Home
  • 442306 - 2,4,6-Trimethylphenol

EMAIL THIS PAGE TO A FRIEND
442306 Supelco

2,4,6-Trimethylphenol

analytical standard

Synonym: Mesitol

  • CAS Number 527-60-6

  • Linear Formula (CH3)3C6H2OH

  • Molecular Weight 136.19

  •  Beilstein Registry Number 1859675

  •  EC Number 208-419-2

  •  MDL number MFCD00002235

  •  PubChem Substance ID 24867791

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   BPRYUXCVCCNUFE-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   220 °C(lit.)
mp   70-72 °C (lit.)
  71-74 °C (lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2430 - class 8 - PG 3 - EHS - Alkylphenols, solid, n.o.s., HI: A,S
WGK Germany 
2
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, PAGE, Pharmaceutical, Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?