EMAIL THIS PAGE TO A FRIEND
442390 Supelco

3-Methylpentane

analytical standard

  • CAS Number 96-14-0

  • Linear Formula (CH3CH2)2CHCH3

  • Molecular Weight 86.18

  •  Beilstein/REAXYS Number 1730734

  •  EC Number 202-481-4

  •  MDL number MFCD00009342

  •  PubChem Substance ID 24867825

Purchase

Properties

grade   analytical standard
vapor density   2.97 (vs air)
vapor pressure   135 mmHg ( 17 °C)
form   neat
autoignition temp.   532 °F
expl. lim.   ~7.7 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.376 (lit.)
bp   64 °C (lit.)
density   0.664 g/mL at 25 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Environmental
Flavors and Fragrances
Food and Beverages
Personal Care
Petroleum
format   neat
storage temp.   room temp
SMILES string   CCC(C)CC
InChI   1S/C6H14/c1-4-6(3)5-2/h6H,4-5H2,1-3H3
InChI key   PFEOZHBOMNWTJB-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes
WGK Germany 
2
Flash Point(F) 
19.4 °F
Flash Point(C) 
-7 °C
Protocols & Articles

Articles

ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2

This application demonstrates the suitability of the Petrocol column for the efficient analysis of hydrocarbons as listed in ASTM method D5134.
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Separation

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q™ PLOT after SPME using a 75 μm PDMS/Carboxen Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q™ PLOT after SPME using a 75 μm PDMS/Carboxen Fiber
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Separation, Solid phase microextractions

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Reliable Identification of F&F Components by GC/MS and Linear Retention Indices and Multidimensional Chromatography [VIDEO]

In this on-demand webinar by Prof. Luigi Mondello of the University of Messina, Italy, highlights the shortcomings of GC/MS analysis in the identification of F&F compounds when using a straightforwar...
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?