• USA Home
  • 442683 - 4-Nitroquinoline N-oxide

EMAIL THIS PAGE TO A FRIEND
442683 Supelco

4-Nitroquinoline N-oxide

analytical standard

Synonym: 4-NQO, 4-Nitroquinoline 1-oxide

  • CAS Number 56-57-5

  • Empirical Formula (Hill Notation) C9H6N2O3

  • Molecular Weight 190.16

  •  Beilstein/REAXYS Number 165756

  •  EC Number 200-281-1

  •  MDL number MFCD00006738

  •  PubChem Substance ID 24867959

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   154-156 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp
SMILES string   [O-][N+](=O)c1cc[n+]([O-])c2ccccc12
InChI   1S/C9H6N2O3/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H
InChI key   YHQDZJICGQWFHK-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?