EMAIL THIS PAGE TO A FRIEND
442697 Supelco

Octadecane

analytical standard

  • CAS Number 593-45-3

  • Linear Formula CH3(CH2)16CH3

  • Molecular Weight 254.49

  •  Beilstein/REAXYS Number 1740138

  •  EC Number 209-790-3

  •  MDL number MFCD00009007

  •  PubChem Substance ID 24867966

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor density   8.8 (vs air)
vapor pressure   1 mmHg ( 119 °C)
description   This product has been discontinued. It has been replaced by product 74691.
form   neat
autoignition temp.   455 °F
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   317 °C (lit.)
mp   26-29 °C (lit.)
density   0.777 g/mL at 25 °C (lit.)
Featured Industry   Petroleum
format   neat
storage temp.   room temp
SMILES string   CCCCCCCCCCCCCCCCCC
InChI   1S/C18H38/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3-18H2,1-2H3
InChI key   RZJRJXONCZWCBN-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
330.8 °F
Flash Point(C) 
166 °C
Protocols & Articles

Articles

GC Analysis of QA Test Mix on SLB®-IL60 before/after Carrier Gas Flow Stopped

From our library of Articles, Sigma-Aldrich presents GC Analysis of QA Test Mix on SLB®-IL60 before/after Carrier Gas Flow Stopped
Keywords: Chromatography, Flame ionization detector, Gas chromatography

GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column

From our library of Articles, Sigma-Aldrich presents GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?