EMAIL THIS PAGE TO A FRIEND
442707 Supelco

Tetracosane

analytical standard

  • CAS Number 646-31-1

  • Linear Formula CH3(CH2)22CH3

  • Molecular Weight 338.65

  •  Beilstein/REAXYS Number 1758462

  •  EC Number 211-474-5

  •  MDL number MFCD00009352

  •  PubChem Substance ID 24867972

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   391 °C (lit.)
mp   49-52 °C (lit.)
Featured Industry   Environmental
Petroleum
format   neat
storage temp.   room temp
SMILES string   CCCCCCCCCCCCCCCCCCCCCCCC
InChI   1S/C24H50/c1-3-5-7-9-11-13-15-17-19-21-23-24-22-20-18-16-14-12-10-8-6-4-2/h3-24H2,1-2H3
InChI key   POOSGDOYLQNASK-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles

Articles

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Total Petroleum Hydrocarbons (TPH) on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

87089 Tetracosane, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?