EMAIL THIS PAGE TO A FRIEND
442830 Supelco

Triphenylene

analytical standard

Synonym: 9,10-Benzophenanthrene

  • CAS Number 217-59-4

  • Empirical Formula (Hill Notation) C18H12

  • Molecular Weight 228.29

  •  Beilstein/REAXYS Number 1342908

  •  EC Number 205-922-9

  •  MDL number MFCD00001108

  •  PubChem Substance ID 24868017

Purchase

Properties

Related Categories Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   SLGBZMMZGDRARJ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   438 °C (lit.)
mp   195-198 °C (lit.)
Featured Industry   Environmental
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
YK2925000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?