• USA Home
  • 48693 - 4-Chloro-3-methylphenol solution

EMAIL THIS PAGE TO A FRIEND
48693 Supelco

4-Chloro-3-methylphenol solution

certified reference material, 500 μg/mL in methanol

Synonym: 4-Chloro-m-cresol

  • CAS Number 59-50-7

  • Empirical Formula (Hill Notation) C7H7ClO

  • Molecular Weight 142.58

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
Quality Level   100
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   500 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   room temp
InChI   1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3
InChI key   CFKMVGJGLGKFKI-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography, Solid phase microextractions

HPLC Analysis of Phenols on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenols on SUPELCOSIL™ LC-8
Keywords: Chromatography, High performance liquid chromatography

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8041: GC Analysis of Phenols on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?