• USA Home
  • 48699 - Chlordane (mixture of isomers)

EMAIL THIS PAGE TO A FRIEND
48699 Supelco

Chlordane (mixture of isomers)

20 μg/mL in methanol, analytical standard

Purchase

Properties

grade   analytical standard
description   technical mixture
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   20 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP

From our library of Articles, Sigma-Aldrich presents GC Analysis of cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of α-Hexachlorocyclohexane (α-HCH) and cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP

From our library of Articles, Sigma-Aldrich presents GC Analysis of α-Hexachlorocyclohexane (α-HCH) and cis-Chlordane Enantiomers on Astec® CHIRALDEX® G-BP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pesticides, Purification

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System - Liver Endocrine
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Endocrine System
Keywords: Cancer, Carcinogens, Respiratory

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?