• USA Home
  • 48721-U - 2-Fluoronaphthalene solution

EMAIL THIS PAGE TO A FRIEND
48721-U Supelco

2-Fluoronaphthalene solution

2000 μg/mL in methylene chloride, analytical standard

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type internal/surrogate
packaging   ampule of 1 mL
concentration   2000 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 625
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?