• USA Home
  • 53552-U - Ascentis® Express Peptide ES-C18, 2.7 Micron Capillary HPLC Column

53552-U Supelco

Ascentis® Express Peptide ES-C18, 2.7 Micron Capillary HPLC Column

L × I.D. 15 cm × 100 μm, 2.7 μm particle size



Related Categories Analytical/Chromatography, Ascentis Express Columns for Fast HPLC, Ascentis Express Peptide ES-C18, 2.7μm HPLC Columns, C18 for Biopolymers, Capillary Dimensions,
feature   endcapped: no
parameter   ≤100 °C temp. range
application(s)   HPLC: suitable
L × I.D.   15 cm × 100 μm
impurities   <5 ppm metals
matrix   Fused-Core particle platform
matrix active group   C18 (octadecyl) phase
particle size   2.7 μm
pore size   160 Å
pH-range   1 - 9
suitability   suitable for L1 per USP
mode of chromatography   reversed phase


Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

LC-MS Grade Solvents and Reagents

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

NONH for all modes of transport
WGK Germany 
Protocols & Articles


Ascentis Express UHPLC Columns with 2.0 μm Fused-Core Particles

The new Supelco Ascentis® Express column line has been designed specifically for UHPLC instruments with 2.0 µm Fused-Core® silica, as diagrammed in Figure 1. Ascentis Express 2.0 µm UHPLC columns are...
Richard A. Henry, Gaurang Parmar
Reporter US, Volume 32.2
Keywords: High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

Related Content

Selection Guide for Ascentis® Express HPLC Columns

Ascentis Express C18 is the first choice for starting a new method. However, when a C18 doesn't give the desired separation or your sample contains compounds that are known to be difficult to retain ...
Keywords: High performance liquid chromatography, Separation, Supercritical fluid chromatography

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?