• USA Home
  • 55018-U - Supelco® Mobile Phase Degassing System
EMAIL THIS PAGE TO A FRIEND
55018-U Supelco
Supelco® Mobile Phase Degassing System
Purchase
Description
General description
• 4 Channel
• 0-5 ml/min Flow Range
• Validation output satisfies system compliance
• Smart sensor detects leaks and communicates with vacuum pump
• LED indicates degassing status
• Unprecedented 4 year warranty
The Supelco Mobile Phase Degassing system with its smart sensor not only detects and alerts you to leaks, it also communicates with the vacuum pump. If a change in vacuum is detected due to mobile phase flow rate changes, the pump can compensate by changing its speed. The validation output records vacuum level to satisfy system compliance and validation requirements. The degassing system has a PTFE AF membrane, with NO-OX fittings and tubing.
Legal Information
NO-OX is a trademark of Systec, Inc.
Supelco is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Milli-Q® Water Purification Solutions
Protocols & Articles
Articles
HPLC Troubleshooting Guide
Although HPLC method development has been improved by advances in column technology and instrumentation, problems still arise. In this guide we offer you a systematic means of isolating, identifying,...
Keywords: Buffers, Capillary electrophoresis, Chromatography, Degradations, Detection methods, Diagnostic, Electronics, Eliminations, Filtration, Fluorescence detector, High performance liquid chromatography, Ion Exchange, Precipitation, Sample preparations, Separation, Solvents
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?