• USA Home
  • 57009-U - Supelclean SPE Method Development Kit

EMAIL THIS PAGE TO A FRIEND
57009-U Supelco

Supelclean SPE Method Development Kit

Kit B

  •  NACRES NB.21

Purchase

Properties

Related Categories Analytical/Chromatography, SPE Method Development Plates and Kits, Sample Preparation & Purification, Solid Phase Extraction (SPE)
Quality Level   100
material   polypropylene tube
Featured Industry   Food and Beverages

Description

General description

Supelclean SPE Method Development Kits consist of an assortment of SPE phase chemistries and cartridge configurations ideal for SPE method development. The range of phase chemistries available for each kit allows the user to profile for compound retention, elution and sample matrix selectivity.

Components

Kit contains 12 of each phase chemistry (100mg/1mL):
• Supelclean LC-18
• Supelclean LC-8
• Supelclean LC-CN
• Supelclean LC-Diol
• Supelclean LC-NH2
• Supelclean LC-Ph
• Supelclean LC-SAX
• Supelclean LC-SCX
• Supelclean LC-WCX
• Supelclean LC-Si

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

SPE Cartridges (Tubes) - Phase Selection Guide

The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions

Solid Phase Extraction: Fundamentals and Recent Developments [Webinar]

Solid Phase Extraction (SPE) has become a popular technique for sample preparation due to its convenience, effectiveness, relatively low solvent usage and versatility. The focus of this webinar is on...
Keywords: Sample preparations, Solid phase extractions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?