• USA Home
  • 57019 - Supelclean SPE Method Development Kit
EMAIL THIS PAGE TO A FRIEND
57019 Supelco
Supelclean SPE Method Development Kit
Kit A
  •  eCl@ss 32161000
Purchase
Properties
Related Categories Analytical/Chromatography, SPE Method Development Plates and Kits, Sample Preparation & Purification, Solid Phase Extraction (SPE)
Quality Level   100
material   polypropylene tube
Featured Industry   Food and Beverages
Description
General description
Supelclean SPE Method Development Kits consist of an assortment of SPE phase chemistries and cartridge configurations ideal for SPE method development. The range of phase chemistries available for each kit allows the user to profile for compound retention, elution and sample matrix selectivity.
Components
Kit contains 6 of each phase chemistry (500mg/3mL):
• Supelclean LC-18
• Supelclean LC-8
• Supelclean LC-CN
• Supelclean LC-Diol
• Supelclean LC-NH2
• Supelclean LC-Ph
• Supelclean LC-SAX
• Supelclean LC-SCX
• Supelclean LC-WCX
• Supelclean LC-Si
Legal Information
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
SPE Cartridges (Tubes) - Phase Selection Guide
The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions
Solid Phase Extraction: Fundamentals and Recent Developments [Webinar]
Solid Phase Extraction (SPE) has become a popular technique for sample preparation due to its convenience, effectiveness, relatively low solvent usage and versatility. The focus of this webinar is on...
Keywords: Sample preparations, Solid phase extractions
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?