• USA Home
  • 57432 - Supelco® Solvent Recovery System
EMAIL THIS PAGE TO A FRIEND
57432 Supelco
Supelco® Solvent Recovery System
SRS-3000, autoclean
Synonym: SRS-3000 SOLVENT RECOVERY SYS. W/AUTOCLEAN
Purchase
Properties
AC/DC input   90 / 240 AC, 50-60
feature   autoclean
mfr. no.   SRS-3000
Description
General description
Supelco Solvent Recovery Systems Recover and reuse clean mobile phase, dispose of only contaminated mobile phase.
• Reduce solvent purchase and disposal costs
• Save money, mobile phase preparation time, and the environment
Supelco SRS-3000 and SRS-1000 Solvent Recovery Systems can save money and time in any isocratic analysis. A microprocessor-controlled solvent switching valve monitors detector output and directs the solvent to the waste reservoir only when a peak is detected (Figure A). When the baseline falls below the threshold you select, the uncontaminated solvent is directed back to the mobile phase reservoir. In a typical isocratic analysis, 80-90% of the mobile phase is uncontaminated and can be recycled (Figure B). Settings for threshold, detection range, and delay time enable you to precisely control the switching valve.
In addition to the basic features mentioned above, the SRS-3000 unit offers validation output (included), an Autoclean option (see below), and storage for up to 10 method files. The validation output provides a continuous, auditable data trail of the solvent recycling valve position, for GMP, GLP, or ISO-9000 protocols. The valve position is recorded by superimposing tick marks over separate copy of the chromatographic signal.
Autoclean Valve - The SRS-3000 system also is available with valve that enables you to select a different solvent to flush the HPLC system. The Autoclean valve is especially useful if you are using a single pump with mobile phases containing buffer or other salts. The Autoclean valve installs between the mobile phase reservoir and the pump. It has two inlet lines, one for the mobile phase and one for the wash solvent. The valve can be factory installed, or you can order it separately and install it yourself.
Economy-Priced Unit
- The economically priced SRS-1000 includes the same solvent-saving features as the SRS-3000 unit. A simpler display and no advanced features (no validation output, Autoclean option, or method storage memory) allow us to keep the price substantially lower.
Both Systems are Ready to Use
- Both systems include a control unit with switching valve, a power cord, a 2-lead signal cable (+/-), PTFE tubing and fittings, and an instruction manual.
In addition to these components, the SRS-3000 system with the Autoclean valve has the wash valve, additional tubing and fittings, a wash start cable, and a pump remote stop cable.
The SRS-3000 and SRS-1000 units meet all CE requirements.
The SRS-1000 units also meet UL and CSA requirements.
Supelco® solvent recovery system is an reliable, accurate and easier to use solvent recycling system used for the recycling of mobile phase used in isocratic high performance liquid chromatographic analysis.
Legal Information
Supelco is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Articles
HPLC Troubleshooting Guide
Although HPLC method development has been improved by advances in column technology and instrumentation, problems still arise. In this guide we offer you a systematic means of isolating, identifying,...
Keywords: Buffers, Capillary electrophoresis, Chromatography, Degradations, Detection methods, Diagnostic, Electronics, Eliminations, Filtration, Fluorescence detector, High performance liquid chromatography, Ion Exchange, Precipitation, Sample preparations, Separation, Solvents
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?