• USA Home
  • 57676-U - Isolation Technologies Precolumn Filter

EMAIL THIS PAGE TO A FRIEND
57676-U Supelco

Isolation Technologies Precolumn Filter

use to protect 2.1 mm I.D. HPLC column, frit diam. 1.5 mm

Synonym: Isolation Technologies Precolumn Filter, 1.5mm Frit

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Inlet Line Filters, Isolation Technologies pre-column filters & frits More...
frit diam.   1.5 mm
compatibility   use to protect 2.1 mm I.D. HPLC column

Description

General description

In-line installation. High capacity inlet filter, with 0.5 μm stainless steel frit. Minimizes dead volume and band broadening, to prevent loss of column efficiency while protecting your column. Includes two each of tubing, nut, and ferrule.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

HPLC Troubleshooting Guide

Although HPLC method development has been improved by advances in column technology and instrumentation, problems still arise. In this guide we offer you a systematic means of isolating, identifying,...
Keywords: Buffers, Capillary electrophoresis, Chromatography, Degradations, Detection methods, Diagnostic, Electronics, Eliminations, Filtration, Fluorescence detector, High performance liquid chromatography, Ion Exchange, Precipitation, Sample preparations, Separation, Solvents

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

accessory

Product #

Description

Add to Cart

57678 Frits for Isolation Technologies Precolumn Filter, frit diam. 1.5 mm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?