• USA Home
  • 57678 - Frits for Isolation Technologies Precolumn Filter
EMAIL THIS PAGE TO A FRIEND
57678 Supelco
Frits for Isolation Technologies Precolumn Filter
frit diam. 1.5 mm
Synonym: Replacement Frit, 1.5mm, for IT Precolumn Filter, 5 pk
Purchase
Properties
Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Inlet Line Filters, Isolation Technologies pre-column filters & frits More...
material   stainless steel frit
packaging   pkg of 10 ea
frit diam.   1.5 mm
pore size   0.5 μm
compatibility   for use with Isolation Technology precolumn filter (57676-U)
Description
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Milli-Q® Water Purification Solutions
Protocols & Articles
Articles
HPLC Troubleshooting Guide
Although HPLC method development has been improved by advances in column technology and instrumentation, problems still arise. In this guide we offer you a systematic means of isolating, identifying,...
Keywords: Buffers, Capillary electrophoresis, Chromatography, Degradations, Detection methods, Diagnostic, Electronics, Eliminations, Filtration, Fluorescence detector, High performance liquid chromatography, Ion Exchange, Precipitation, Sample preparations, Separation, Solvents
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?