• USA Home
  • MET12159B - Pentachlorothiophenol

EMAIL THIS PAGE TO A FRIEND
MET12159B Supelco

Pentachlorothiophenol

analytical standard

Synonym: Pentachlorobenzenethiol

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., MET-690B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   223-227 °C (lit.)
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   Sc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
InChI   1S/C6HCl5S/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
InChI key   LLMLGZUZTFMXSA-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3335
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?