• USA Home
  • MET690B - Pentachlorothiophenol

EMAIL THIS PAGE TO A FRIEND
MET690B Supelco

Pentachlorothiophenol

analytical standard

Synonym: Pentachlorobenzenethiol

Purchase

Properties

grade   analytical standard
InChI Key   LLMLGZUZTFMXSA-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., MET-690B
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   223-227 °C (lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3335
WGK Germany 
3
RTECS 
DC1925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?