EMAIL THIS PAGE TO A FRIEND
PS2057 Supelco

Flutolanil

analytical standard

Purchase

Properties

Related Categories Additional Standards, All Fungicides, Alpha Index of All Pesticide Standards, Amide structure, Analytical Standards,
grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-2057
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
CV5581320

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 2) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 2) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?