EMAIL THIS PAGE TO A FRIEND
PS220 Supelco

Nabam

analytical standard

Synonym: Disodium ethylenebis(dithiocarbamate)

Purchase

Properties

grade   analytical standard
InChI Key   UQJQVUOTMVCFHX-UHFFFAOYSA-L
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-220
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
FA6825000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?