• USA Home
  • PS562 - 1,2,4-Tributyl phosphorotrithioate

EMAIL THIS PAGE TO A FRIEND
PS562 Supelco

1,2,4-Tributyl phosphorotrithioate

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-562
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
TG5425000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?