EMAIL THIS PAGE TO A FRIEND
PS6 Supelco

Dichlorophen

analytical standard

Synonym: Bis(5-chloro-2-hydroxyphenyl)methane, Dichlorophen, Dichlorophene

  • CAS Number 97-23-4

  • Linear Formula CH2[C6H3(Cl)OH]2

  • Molecular Weight 269.12

  •  Beilstein Registry Number 1884514

  •  EC Number 202-567-1

  •  MDL number MFCD00002322

  •  PubChem Substance ID 24899246

Purchase

Properties

grade   analytical standard
InChI Key   MDNWOSOZYLHTCG-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-6
mp   168-172 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
SM0175000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?