EMAIL THIS PAGE TO A FRIEND
PS962 Supelco

MGK 264

analytical standard

Synonym: N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide

Purchase

Properties

grade   analytical standard
InChI Key   WLLGXSLBOPFWQV-OTHKPKEBSA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-962
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
RTECS 
RB8575000
Flash Point(F) 
350.6 °F
Flash Point(C) 
177 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?