• USA Home
  • 67288 - Alkaline earth metal mix for ICP

EMAIL THIS PAGE TO A FRIEND
67288 Supelco

Alkaline earth metal mix for ICP

TraceCERT®, 5 elements, 100 mg/L in nitric acid

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   certified reference material
  TraceCERT®
shelf life   limited shelf life, expiry date on the label
concentration   100 mg/L in nitric acid
application(s)   AAS: suitable
  ICP: suitable
format   multi-component solution

Description

Analysis Note

100 mg/L each: Be, Mg, Ca, Sr and Ba in 2% nitric acid

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?