• USA Home
  • 19680 - Aminoallyl-dUTP-Atto 425 triethylammonium salt solution

EMAIL THIS PAGE TO A FRIEND
19680 Sigma-Aldrich

Aminoallyl-dUTP-Atto 425 triethylammonium salt solution

≥95.0% (HPLC)

Synonym: 5-(3-Aminoallyl)-2′-deoxyuridine 5′-triphosphate labeled with Atto 425 triethylammonium salt, Atto 425-aminoallyl-dUTP

  • Empirical Formula (Hill Notation) C34H45N4O19P3 · xC6H15N

  • Molecular Weight 906.66 (free acid basis)

  •  PubChem Substance ID 329751929

  •  NACRES NA.32

Purchase

Properties

Quality Level   100
assay   ≥95.0% (HPLC)
packaging   pkg of 10 μL
concentration   1 mM
pH   7.0-8.0
storage temp.   −20°C
SMILES string   OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](O)C[C@H](N2C(NC(C(/C=C/CNC(CCCN3C(C=C(OC(C(C(OCC)=O)=C4)=O)C4=C5)=C5C(C)CC3(C)C)=O)=C2)=O)=O)O1)=O)=O)(O)=O.CCN(CC)CC
InChI   1S/C34H45N4O19P3.C6H15N/c1-5-52-31(42)23-13-21-12-22-19(2)16-34(3,4)38(24(22)14-26(21)55-32(23)43)11-7-9-28(40)35-10-6-8-20-17-37(33(44)36-30(20)41)29-15-25(39)27(54-29)18-53-59(48,49)57-60(50,51)56-58(45,46)47;1-4-7(5-2)6-3/h6,8,12-14,17,19,25,27,29,39H,5,7,9-11,15-16,18H2,1-4H3,(H,35,40)(H,48,49)(H,50,51)(H,36,41,44)(H2,45,46,47);4-6H2,1-3H3/b8-6+;/t19?,25-,27-,29-;/m1./s1
InChI key   SFGJWXZCNJBQKY-FISLXKFISA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?