• USA Home
  • 30387 - Aminoallyl-dUTP-Atto 550 triethylammonium salt solution

EMAIL THIS PAGE TO A FRIEND
30387 Sigma-Aldrich

Aminoallyl-dUTP-Atto 550 triethylammonium salt solution

≥95.0% (HPLC)

Synonym: 5-(3-Aminoallyl)-2′-deoxyuridine 5′-triphosphate labeled with Atto 550 triethylammonium salt, Atto 550-aminoallyl-dUTP

  •  PubChem Substance ID 329753490

  •  NACRES NA.32

Purchase

Properties

Quality Level   100
assay   ≥95.0% (HPLC)
mol wt   Mw 1099.22 g/mol (free acid)
pH   7.0-8.0
storage temp.   −20°C
SMILES string   OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](O)C[C@H](N2C(NC(C(/C=C/CNC)=C2)=O)=O)O1)=O)=O)(O)=O.CCN(CC)CC
InChI   1S/C13H22N3O14P3.C6H15N/c1-14-4-2-3-8-6-16(13(19)15-12(8)18)11-5-9(17)10(28-11)7-27-32(23,24)30-33(25,26)29-31(20,21)22;1-4-7(5-2)6-3/h2-3,6,9-11,14,17H,4-5,7H2,1H3,(H,23,24)(H,25,26)(H,15,18,19)(H2,20,21,22);4-6H2,1-3H3/b3-2+;/t9-,10-,11-;/m1./s1
InChI key   VPWSAISNRHVMRE-HZRKYXBQSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?