• USA Home
  • 69303 - Aminoallyl-XX-dUTP-Atto 488 triethylammonium salt solution

EMAIL THIS PAGE TO A FRIEND
69303 Sigma-Aldrich

Aminoallyl-XX-dUTP-Atto 488 triethylammonium salt solution

≥95.0% (HPLC)

Synonym: 5-(3-Aminoallyl)-2′-deoxyuridine 5′-triphosphate labeled with Atto 488 triethylammonium salt

  • Empirical Formula (Hill Notation) C49H63N8O25P3S2 · xC6H15N

  • Molecular Weight 1321.11 (free acid basis)

  •  PubChem Substance ID 329761555

Purchase

Properties

Quality Level   100
assay   ≥95.0% (HPLC)
pH   7.0-8.0
storage temp.   −20°C
SMILES string   OP(OP(O)(OP(O)(OC[C@@H]1[C@@H](O)C[C@H](N2C(NC(C(/C=C/CNC(CCCCCNC(CCCCCNC(CCCN(C)C(C3=CC=CC=C3C4=C(C=C5)C(OC6=C4C=CC(N)=C6S(=O)(O)=O)=C(S(=O)([O-])=O)C5=[NH2+])=O)=O)=O)=O)=C2)=O)=O)O1)=O)=O)(O)=O.CCN(CC)CC
InChI   1S/C49H63N8O25P3S2.C6H15N/c1-56(48(63)31-14-7-6-13-30(31)42-32-18-20-34(50)45(86(72,73)74)43(32)80-44-33(42)19-21-35(51)46(44)87(75,76)77)25-11-17-40(61)53-23-9-2-4-15-38(59)52-22-8-3-5-16-39(60)54-24-10-12-29-27-57(49(64)55-47(29)62)41-26-36(58)37(79-41)28-78-84(68,69)82-85(70,71)81-83(65,66)67;1-4-7(5-2)6-3/h6-7,10,12-14,18-21,27,36-37,41,50,58H,2-5,8-9,11,15-17,22-26,28,51H2,1H3,(H,52,59)(H,53,61)(H,54,60)(H,68,69)(H,70,71)(H,55,62,64)(H2,65,66,67)(H,72,73,74)(H,75,76,77);4-6H2,1-3H3/b12-10+,50-34?;/t36-,37-,41-;/m1./s1
InChI key   MSPMRAHPYLWHQF-NCFMSHCKSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?