• USA Home
  • 69383 - Aminoallyl-dUTP-Texas Red® triethylammonium salt solution

EMAIL THIS PAGE TO A FRIEND
69383 Sigma-Aldrich

Aminoallyl-dUTP-Texas Red® triethylammonium salt solution

≥95.0% (HPLC)

Synonym: 5-(3-Aminoallyl)-2′-deoxyuridine 5′-triphosphate, labeled with Texas Red® triethylammonium salt

  • Empirical Formula (Hill Notation) C49H59N6O21P3S2 · xC6H15N

  • Molecular Weight 1225.07 (free acid basis)

  •  NACRES NA.32

Purchase

Properties

Quality Level   100
assay   ≥95.0% (HPLC)
pH   7.0-8.0
storage temp.   −20°C

Description

Legal Information

Texas Red is a registered trademark of Life Technologies

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?