EMAIL THIS PAGE TO A FRIEND
1223209 USP

Allyl disulfide

United States Pharmacopeia (USP) Reference Standard

Synonym: Diallyl disulfide

  • CAS Number 2179-57-9

  • Linear Formula CH2=CHCH2SSCH2CH=CH2

  • Molecular Weight 146.27

  •  Beilstein/REAXYS Number 1699241

  •  MDL number MFCD00008656

  •  PubChem Substance ID 329749658

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards A - B More...
grade   pharmaceutical primary standard
vapor density   >5 (vs air)
vapor pressure   1 mmHg ( 20 °C)
mfr. no.   USP
refractive index   n20/D 1.541 (lit.)
bp   180-195 °C (lit.)
density   1.008 g/mL at 25 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
SMILES string   C=CCSSCC=C
InChI   1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2
InChI key   PFRGXCVKLLPLIP-UHFFFAOYSA-N

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Organosulfur compound that is naturally found in garlic. Contributes to the pungent odor of crushed garlic. Anticancer agent and may provide protection against cardiovascular disease.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 1992 6.1(3) / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
143.6 °F - closed cup
Flash Point(C) 
62 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?