EMAIL THIS PAGE TO A FRIEND
1231207 USP

Ebastine

United States Pharmacopeia (USP) Reference Standard

Synonym: 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(diphenylmethoxy)-1-piperidinyl]-1-butanone

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards E - F More...
InChI Key   MJJALKDDGIKVBE-UHFFFAOYSA-N
Featured Industry   Pharmaceutical (small molecule)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Ebastine is a non-sedating histamine H1 receptor antagonist, which inhibits allergen-induced bronchospasm in conscious guinea pigs. Unlike other compounds in this category, ebastine does not prolong the QT interval at up to five times the recommended therapeutic dose.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Hazard statements 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?