• USA Home
  • 1231626 - Ecamsule Related Compound B

EMAIL THIS PAGE TO A FRIEND
1231626 USP

Ecamsule Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym: 4-(Hydroxymethyl)benzoic acid

  • CAS Number 3006-96-0

  • Linear Formula HOCH2C6H4CO2H

  • Molecular Weight 152.15

  •  Beilstein/REAXYS Number 2690023

  •  MDL number MFCD00017598

  •  PubChem Substance ID 329749703

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards E - F More...
mp   182-185 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
SMILES string   OCc1ccc(cc1)C(O)=O
InChI   1S/C8H8O3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,9H,5H2,(H,10,11)
InChI key   WWYFPDXEIFBNKE-UHFFFAOYSA-N

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?