EMAIL THIS PAGE TO A FRIEND
V000375 Sigma-Aldrich

L-Aspartic acid

99.0%

Synonym: (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

  • CAS Number 56-84-8

  • Linear Formula HO2CCH2CH(NH2)CO2H

  • Molecular Weight 133.10

  •  Beilstein/REAXYS Number 1723530

  •  EC Number 200-291-6

  •  MDL number MFCD00002616

  •  PubChem Substance ID 329827965

Purchase

Properties

assay   99.0%
form   powder
color   white
mp   >300 °C (dec.) (lit.)
SMILES string   N[C@@H](CC(O)=O)C(O)=O
InChI   1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1
InChI key   CKLJMWTZIZZHCS-REOHCLBHSA-N

Description

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
CI9098500
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?