EMAIL THIS PAGE TO A FRIEND
V002030 Sigma-Aldrich

1-Butanol

UV HPLC spectroscopic, 99.5%

Synonym: n-Butanol, Butyl alcohol

Purchase

Properties

grade   UV HPLC spectroscopic
vapor density   2.55 (vs air)
assay   99.5%
autoignition temp.   649 °F
expl. lim.   11.2 %
refractive index   n20/D 1.399 (lit.)
bp   116-118 °C (lit.)
mp   −90 °C (lit.)
density   0.81 g/mL at 25 °C (lit.)
SMILES string   CCCCO
InChI   1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3
InChI key   LRHPLDYGYMQRHN-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1120 - class 3 - PG 3 - Butanols
WGK Germany 
WGK 1
RTECS 
EO1400000
Flash Point(F) 
95.0 °F - Pensky-Martens closed cup
Flash Point(C) 
35 °C - Pensky-Martens closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?