EMAIL THIS PAGE TO A FRIEND
V003390 Sigma-Aldrich

(−)-Menthol

USP, 97%

Synonym: (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

  • CAS Number 2216-51-5

  • Empirical Formula (Hill Notation) C10H20O

  • Molecular Weight 156.27

  •  Beilstein/REAXYS Number 1902293

  •  EC Number 218-690-9

  •  MDL number MFCD00062979

  •  PubChem Substance ID 329828898

Purchase

Properties

Agency/Method   USP
assay   97%
SMILES string   CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
InChI   1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1
InChI key   NOOLISFMXDJSKH-KXUCPTDWSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?