• USA Home
  • V900031 - Ammonium iron(II) sulfate hexahydrate

EMAIL THIS PAGE TO A FRIEND
V900031 Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

Vetec reagent grade, 98%

Synonym: Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Purchase

Properties

Related Categories Essential Chemicals, Iron Salts, Research Essentials, Salts
grade   Vetec reagent grade
InChI Key   MQLVWQSVRZVNIP-UHFFFAOYSA-L
assay   98%
mp   100 °C (dec.) (lit.)

Description

Packaging

500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?