• USA Home
  • V900096 - Sodium carbonate decahydrate

EMAIL THIS PAGE TO A FRIEND
V900096 Sigma-Aldrich

Sodium carbonate decahydrate

Vetec, reagent grade, 99%

Synonym: Sal soda, Soda

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
grade   reagent grade
product line   Vetec
assay   99%
form   crystals
pH   11.0-12.0 (25 °C, 50 g/L)
SMILES string   O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
InChI   1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2
InChI key   XYQRXRFVKUPBQN-UHFFFAOYSA-L

Description

Packaging

500, 6×500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?