• USA Home
  • V900123 - Cyclopentyl methyl ether

EMAIL THIS PAGE TO A FRIEND
V900123 Sigma-Aldrich

Cyclopentyl methyl ether

Vetec, reagent grade, 99%

Synonym: CPME

Purchase

Properties

Related Categories ACS and Reagent Grade Solvents, Reagent Grade Solvents, Solvent by Application, Solvents
grade   reagent grade
product line   Vetec
assay   99%
form   liquid
pH   7-9 (25 °C, 98.2 g/L)
bp   106 °C/760 mmHg
mp   -140 °C (lit.)
density   0.86 g/mL at 25 °C
SMILES string   COC1CCCC1
InChI   1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3
InChI key   SKTCDJAMAYNROS-UHFFFAOYSA-N

Description

Packaging

100 mL in glass bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3271 3 / PGII
WGK Germany 
3
Flash Point(F) 
30.2 °F
Flash Point(C) 
-1 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?