• USA Home
  • V900137 - Zinc carbonate basic

EMAIL THIS PAGE TO A FRIEND
V900137 Sigma-Aldrich

Zinc carbonate basic

Vetec, reagent grade

Synonym: Zinc hydroxide carbonate

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Zinc Salts
grade   reagent grade
product line   Vetec
assay   58.0-61.0% (EDTA titration)
form   powder
SMILES string   [Zn++].[Zn++].O[Zn]O.O[Zn]O.O[Zn]O.[O-]C([O-])=O.[O-]C([O-])=O
InChI   1S/2CH2O3.6H2O.5Zn/c2*2-1(3)4;;;;;;;;;;;/h2*(H2,2,3,4);6*1H2;;;;;/q;;;;;;;;5*+2/p-10
InChI key   UOURRHZRLGCVDA-UHFFFAOYSA-D

Description

Packaging

500, 6×500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?