• USA Home
  • V900154 - Ethylene glycol monopropyl ether

EMAIL THIS PAGE TO A FRIEND
V900154 Sigma-Aldrich

Ethylene glycol monopropyl ether

Vetec, reagent grade, 99%

Synonym: 2-Propoxyethanol, Propyl glycol

  • CAS Number 2807-30-9

  • Linear Formula CH3CH2CH2OCH2CH2OH

  • Molecular Weight 104.15

  •  Beilstein/REAXYS Number 1731983

  •  EC Number 220-548-6

  •  MDL number MFCD00072645

  •  PubChem Substance ID 329829590

Purchase

Properties

Related Categories ACS and Reagent Grade Solvents, Reagent Grade Solvents, Solvent by Application, Solvents
grade   reagent grade
vapor pressure   1.3 mmHg ( 20 °C)
product line   Vetec
assay   99%
form   liquid
impurities   ≤0.1% water
water
refractive index   n20/D 1.413 (lit.)
bp   150-153 °C (lit.)
mp   −75 °C (lit.)
density   0.913 g/mL at 25 °C (lit.)
SMILES string   CCCOCCO
InChI   1S/C5H12O2/c1-2-4-7-5-3-6/h6H,2-5H2,1H3
InChI key   YEYKMVJDLWJFOA-UHFFFAOYSA-N

Description

Packaging

500 mL in glass bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
1
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?